Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 11 - 12 | 1966 | 84 | |
XM_021845079.1 | XP_021700771.1 | 11 - 12 | 1922 | 84 | ||
XM_021845080.1 | XP_021700772.1 | 10 - 11 | 1598 | 84 |
