Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 12 - 13 | 2091 | 75 | |
XM_021845079.1 | XP_021700771.1 | 12 - 13 | 2047 | 75 | ||
XM_021845080.1 | XP_021700772.1 | 11 - 12 | 1723 | 75 |
