Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 15 - 16 | 2732 | 92 | |
XM_021845079.1 | XP_021700771.1 | 15 - 16 | 2688 | 92 | ||
XM_021845080.1 | XP_021700772.1 | 14 - 15 | 2364 | 92 |
