Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 17 - 18 | 3059 | 94 | |
XM_021845079.1 | XP_021700771.1 | 17 - 18 | 3015 | 94 | ||
XM_021845080.1 | XP_021700772.1 | 16 - 17 | 2691 | 94 |
