Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 18 - 19 | 3708 | 108 | |
XM_021845079.1 | XP_021700771.1 | 18 - 19 | 3664 | 108 | ||
XM_021845080.1 | XP_021700772.1 | 17 - 18 | 3340 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
