Assay Details
Gene Symbol: LOC5566347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845078.1 | XP_021700770.1 | 9 - 10 | 1496 | 64 | |
XM_021845079.1 | XP_021700771.1 | 9 - 10 | 1452 | 64 | ||
XM_021845080.1 | XP_021700772.1 | 8 - 9 | 1128 | 64 |

More Information
Set Membership: |
![]() ![]() |
