Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 7 - 8 | 3168 | 88 | |
XM_021844720.1 | XP_021700412.1 | 7 - 8 | 3174 | 88 | ||
XM_021844721.1 | XP_021700413.1 | 7 - 8 | 3153 | 88 | ||
XM_021844722.1 | XP_021700414.1 | 7 - 8 | 3062 | 88 | ||
XM_021844723.1 | XP_021700415.1 | 7 - 8 | 3078 | 88 | ||
XM_021844724.1 | XP_021700416.1 | 7 - 8 | 3072 | 88 | ||
XM_021844725.1 | XP_021700417.1 | 7 - 8 | 3068 | 88 | ||
XM_021844726.1 | XP_021700418.1 | 7 - 8 | 3080 | 88 | ||
XM_021844727.1 | XP_021700419.1 | 7 - 8 | 3047 | 88 | ||
XM_021844728.1 | XP_021700420.1 | 7 - 8 | 3029 | 88 | ||
XM_021844729.1 | XP_021700421.1 | 7 - 8 | 3022 | 88 | ||
XM_021844730.1 | XP_021700422.1 | 7 - 8 | 3131 | 88 | ||
XM_021844731.1 | XP_021700423.1 | 6 - 7 | 2936 | 88 | ||
XM_021844732.1 | XP_021700424.1 | 6 - 7 | 2942 | 88 | ||
XM_021844733.1 | XP_021700425.1 | 6 - 7 | 2926 | 88 | ||
XM_021844734.1 | XP_021700426.1 | 6 - 7 | 2921 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
