Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 6 - 7 | 2516 | 86 | |
XM_021844720.1 | XP_021700412.1 | 6 - 7 | 2522 | 86 | ||
XM_021844721.1 | XP_021700413.1 | 6 - 7 | 2501 | 86 | ||
XM_021844722.1 | XP_021700414.1 | 6 - 7 | 2410 | 86 | ||
XM_021844723.1 | XP_021700415.1 | 6 - 7 | 2426 | 86 | ||
XM_021844724.1 | XP_021700416.1 | 6 - 7 | 2420 | 86 | ||
XM_021844725.1 | XP_021700417.1 | 6 - 7 | 2416 | 86 | ||
XM_021844726.1 | XP_021700418.1 | 6 - 7 | 2428 | 86 | ||
XM_021844727.1 | XP_021700419.1 | 6 - 7 | 2395 | 86 | ||
XM_021844728.1 | XP_021700420.1 | 6 - 7 | 2377 | 86 | ||
XM_021844729.1 | XP_021700421.1 | 6 - 7 | 2370 | 86 | ||
XM_021844730.1 | XP_021700422.1 | 6 - 7 | 2479 | 86 | ||
XM_021844731.1 | XP_021700423.1 | 5 - 6 | 2284 | 86 | ||
XM_021844732.1 | XP_021700424.1 | 5 - 6 | 2290 | 86 | ||
XM_021844733.1 | XP_021700425.1 | 5 - 6 | 2274 | 86 | ||
XM_021844734.1 | XP_021700426.1 | 5 - 6 | 2269 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
