Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 5 - 6 | 2342 | 107 | |
XM_021844720.1 | XP_021700412.1 | 5 - 6 | 2348 | 107 | ||
XM_021844721.1 | XP_021700413.1 | 5 - 6 | 2327 | 107 | ||
XM_021844722.1 | XP_021700414.1 | 5 - 6 | 2236 | 107 | ||
XM_021844723.1 | XP_021700415.1 | 5 - 6 | 2252 | 107 | ||
XM_021844724.1 | XP_021700416.1 | 5 - 6 | 2246 | 107 | ||
XM_021844725.1 | XP_021700417.1 | 5 - 6 | 2242 | 107 | ||
XM_021844726.1 | XP_021700418.1 | 5 - 6 | 2254 | 107 | ||
XM_021844727.1 | XP_021700419.1 | 5 - 6 | 2221 | 107 | ||
XM_021844728.1 | XP_021700420.1 | 5 - 6 | 2203 | 107 | ||
XM_021844729.1 | XP_021700421.1 | 5 - 6 | 2196 | 107 | ||
XM_021844730.1 | XP_021700422.1 | 5 - 6 | 2305 | 107 | ||
XM_021844731.1 | XP_021700423.1 | 4 - 5 | 2110 | 107 | ||
XM_021844732.1 | XP_021700424.1 | 4 - 5 | 2116 | 107 | ||
XM_021844733.1 | XP_021700425.1 | 4 - 5 | 2100 | 107 | ||
XM_021844734.1 | XP_021700426.1 | 4 - 5 | 2095 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
