Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 4 - 5 | 1695 | 66 | |
XM_021844720.1 | XP_021700412.1 | 4 - 5 | 1701 | 66 | ||
XM_021844721.1 | XP_021700413.1 | 4 - 5 | 1680 | 66 | ||
XM_021844722.1 | XP_021700414.1 | 4 - 5 | 1589 | 66 | ||
XM_021844723.1 | XP_021700415.1 | 4 - 5 | 1605 | 66 | ||
XM_021844724.1 | XP_021700416.1 | 4 - 5 | 1599 | 66 | ||
XM_021844725.1 | XP_021700417.1 | 4 - 5 | 1595 | 66 | ||
XM_021844726.1 | XP_021700418.1 | 4 - 5 | 1607 | 66 | ||
XM_021844727.1 | XP_021700419.1 | 4 - 5 | 1574 | 66 | ||
XM_021844728.1 | XP_021700420.1 | 4 - 5 | 1556 | 66 | ||
XM_021844729.1 | XP_021700421.1 | 4 - 5 | 1549 | 66 | ||
XM_021844730.1 | XP_021700422.1 | 4 - 5 | 1658 | 66 | ||
XM_021844731.1 | XP_021700423.1 | 3 - 4 | 1463 | 66 | ||
XM_021844732.1 | XP_021700424.1 | 3 - 4 | 1469 | 66 | ||
XM_021844733.1 | XP_021700425.1 | 3 - 4 | 1453 | 66 | ||
XM_021844734.1 | XP_021700426.1 | 3 - 4 | 1448 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
