Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 3 - 4 | 891 | 66 | |
XM_021844720.1 | XP_021700412.1 | 3 - 4 | 897 | 66 | ||
XM_021844721.1 | XP_021700413.1 | 3 - 4 | 876 | 66 | ||
XM_021844722.1 | XP_021700414.1 | 3 - 4 | 785 | 66 | ||
XM_021844723.1 | XP_021700415.1 | 3 - 4 | 801 | 66 | ||
XM_021844724.1 | XP_021700416.1 | 3 - 4 | 795 | 66 | ||
XM_021844725.1 | XP_021700417.1 | 3 - 4 | 791 | 66 | ||
XM_021844726.1 | XP_021700418.1 | 3 - 4 | 803 | 66 | ||
XM_021844727.1 | XP_021700419.1 | 3 - 4 | 770 | 66 | ||
XM_021844728.1 | XP_021700420.1 | 3 - 4 | 752 | 66 | ||
XM_021844729.1 | XP_021700421.1 | 3 - 4 | 745 | 66 | ||
XM_021844730.1 | XP_021700422.1 | 3 - 4 | 854 | 66 | ||
XM_021844731.1 | XP_021700423.1 | 2 - 3 | 659 | 66 | ||
XM_021844732.1 | XP_021700424.1 | 2 - 3 | 665 | 66 | ||
XM_021844733.1 | XP_021700425.1 | 2 - 3 | 649 | 66 | ||
XM_021844734.1 | XP_021700426.1 | 2 - 3 | 644 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
