Assay Details
Gene Symbol: LOC5571941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844719.1 | XP_021700411.1 | 1 - 2 | 450 | 86 | |
XM_021844730.1 | XP_021700422.1 | 1 - 2 | 450 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021844719.1, XM_021844730.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
