Assay Details
Gene Symbol: LOC5570003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849900.1 | XP_021705592.1 | 4 - 5 | 1461 | 84 | |
XM_021849901.1 | XP_021705593.1 | 4 - 5 | 1443 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
