Assay Details
Gene Symbol: LOC5573278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654440.2 | XP_001654490.2 | 2 - 3 | 317 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5573278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654440.2 | XP_001654490.2 | 2 - 3 | 317 | 74 |
Set Membership: |
![]() ![]() ![]() |