Assay Details
Gene Symbol: LOC5566058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650374.2 | XP_001650424.1 | 4 - 5 | 1816 | 81 | |
XM_021853165.1 | XP_021708857.1 | 3 - 4 | 1019 | 81 | ||
XM_021853166.1 | XP_021708858.1 | 3 - 4 | 1029 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
