Assay Details
Gene Symbol: LOC5566058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650374.2 | XP_001650424.1 | 5 - 6 | 2157 | 111 | |
XM_021853165.1 | XP_021708857.1 | 4 - 5 | 1360 | 111 | ||
XM_021853166.1 | XP_021708858.1 | 4 - 5 | 1370 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
