Assay Details
Gene Symbol: LOC5564807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846410.1 | XP_021702102.1 | 9 - 10 | 2164 | 70 | |
XM_021846411.1 | XP_021702103.1 | 10 - 11 | 1947 | 70 | ||
XM_021846413.1 | XP_021702105.1 | 9 - 10 | 2177 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
