Assay Details
Gene Symbol: LOC5575304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852830.1 | XP_021708522.1 | 3 - 4 | 1128 | 117 | |
XM_021852831.1 | XP_021708523.1 | 3 - 4 | 992 | 117 | ||
XM_021852832.1 | XP_021708524.1 | 2 - 3 | 883 | 117 | ||
XM_021852833.1 | XP_021708525.1 | 2 - 3 | 1800 | 117 | ||
XM_021852834.1 | XP_021708526.1 | 2 - 3 | 1801 | 117 | ||
XM_021852835.1 | XP_021708527.1 | 2 - 3 | 1800 | 117 |

More Information
Set Membership: |
![]() ![]() |
