Assay Details
Gene Symbol: LOC5566742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851195.1 | XP_021706887.1 | 8 - 9 | 2236 | 93 | |
XM_021851196.1 | XP_021706888.1 | 8 - 9 | 2192 | 93 | ||
XM_021851197.1 | XP_021706889.1 | 8 - 9 | 2169 | 93 | ||
XM_021851198.1 | XP_021706890.1 | 8 - 9 | 2113 | 93 | ||
XM_021851199.1 | XP_021706891.1 | 8 - 9 | 2069 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
