Assay Details
Gene Symbol: LOC5566742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851195.1 | XP_021706887.1 | 7 - 8 | 1936 | 83 | |
XM_021851196.1 | XP_021706888.1 | 7 - 8 | 1892 | 83 | ||
XM_021851197.1 | XP_021706889.1 | 7 - 8 | 1869 | 83 | ||
XM_021851198.1 | XP_021706890.1 | 7 - 8 | 1813 | 83 | ||
XM_021851199.1 | XP_021706891.1 | 7 - 8 | 1769 | 83 |
