Assay Details
Gene Symbol: LOC5566742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851195.1 | XP_021706887.1 | 3 - 4 | 1304 | 103 | |
XM_021851196.1 | XP_021706888.1 | 3 - 4 | 1260 | 103 | ||
XM_021851197.1 | XP_021706889.1 | 3 - 4 | 1237 | 103 | ||
XM_021851198.1 | XP_021706890.1 | 3 - 4 | 1181 | 103 | ||
XM_021851199.1 | XP_021706891.1 | 3 - 4 | 1137 | 103 |

More Information
Set Membership: |
![]() ![]() |
