Assay Details
Gene Symbol: LOC5566742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851195.1 | XP_021706887.1 | 2 - 3 | 1154 | 98 | |
XM_021851196.1 | XP_021706888.1 | 2 - 3 | 1110 | 98 | ||
XM_021851197.1 | XP_021706889.1 | 2 - 3 | 1087 | 98 | ||
XM_021851198.1 | XP_021706890.1 | 2 - 3 | 1031 | 98 | ||
XM_021851199.1 | XP_021706891.1 | 2 - 3 | 987 | 98 |
