Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 16 - 17 | 4851 | 69 | |
XM_021847847.1 | XP_021703539.1 | 16 - 17 | 4845 | 69 | ||
XM_021847848.1 | XP_021703540.1 | 16 - 17 | 4588 | 69 | ||
XM_021847849.1 | XP_021703541.1 | 16 - 17 | 4477 | 69 | ||
XM_021847850.1 | XP_021703542.1 | 20 - 21 | 6189 | 69 | ||
XM_021847851.1 | XP_021703543.1 | 19 - 20 | 6051 | 69 | ||
XM_021847852.1 | XP_021703544.1 | 18 - 19 | 5922 | 69 | ||
XM_021847853.1 | XP_021703545.1 | 10 - 11 | 4294 | 69 |

More Information
Set Membership: |
![]() ![]() |
