Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 25 - 26 | 6642 | 63 | |
XM_021847847.1 | XP_021703539.1 | 25 - 26 | 6636 | 63 | ||
XM_021847848.1 | XP_021703540.1 | 25 - 26 | 6379 | 63 | ||
XM_021847849.1 | XP_021703541.1 | 25 - 26 | 6268 | 63 | ||
XM_021847850.1 | XP_021703542.1 | 29 - 30 | 7980 | 63 | ||
XM_021847851.1 | XP_021703543.1 | 28 - 29 | 7842 | 63 | ||
XM_021847852.1 | XP_021703544.1 | 27 - 28 | 7713 | 63 | ||
XM_021847853.1 | XP_021703545.1 | 19 - 20 | 6085 | 63 |

More Information
Set Membership: |
![]() ![]() |
