Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 32 - 33 | 8204 | 99 | |
XM_021847847.1 | XP_021703539.1 | 32 - 33 | 8198 | 99 | ||
XM_021847848.1 | XP_021703540.1 | 32 - 33 | 7941 | 99 | ||
XM_021847849.1 | XP_021703541.1 | 32 - 33 | 7830 | 99 | ||
XM_021847850.1 | XP_021703542.1 | 36 - 37 | 9542 | 99 | ||
XM_021847851.1 | XP_021703543.1 | 35 - 36 | 9404 | 99 | ||
XM_021847852.1 | XP_021703544.1 | 34 - 35 | 9275 | 99 | ||
XM_021847853.1 | XP_021703545.1 | 26 - 27 | 7647 | 99 |
