Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 34 - 35 | 9622 | 63 | |
XM_021847847.1 | XP_021703539.1 | 34 - 35 | 9616 | 63 | ||
XM_021847848.1 | XP_021703540.1 | 34 - 35 | 9359 | 63 | ||
XM_021847849.1 | XP_021703541.1 | 34 - 35 | 9248 | 63 | ||
XM_021847850.1 | XP_021703542.1 | 38 - 39 | 10960 | 63 | ||
XM_021847851.1 | XP_021703543.1 | 37 - 38 | 10822 | 63 | ||
XM_021847852.1 | XP_021703544.1 | 36 - 37 | 10693 | 63 | ||
XM_021847853.1 | XP_021703545.1 | 28 - 29 | 9065 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
