Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 4 - 5 | 803 | 87 | |
XM_021847847.1 | XP_021703539.1 | 4 - 5 | 797 | 87 | ||
XM_021847848.1 | XP_021703540.1 | 4 - 5 | 540 | 87 | ||
XM_021847849.1 | XP_021703541.1 | 4 - 5 | 429 | 87 | ||
XM_021847850.1 | XP_021703542.1 | 8 - 9 | 2159 | 87 | ||
XM_021847851.1 | XP_021703543.1 | 8 - 9 | 2159 | 87 | ||
XM_021847852.1 | XP_021703544.1 | 8 - 9 | 2159 | 87 |
