Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 5 - 6 | 1320 | 95 | |
XM_021847847.1 | XP_021703539.1 | 5 - 6 | 1314 | 95 | ||
XM_021847848.1 | XP_021703540.1 | 5 - 6 | 1057 | 95 | ||
XM_021847849.1 | XP_021703541.1 | 5 - 6 | 946 | 95 | ||
XM_021847850.1 | XP_021703542.1 | 9 - 10 | 2676 | 95 | ||
XM_021847851.1 | XP_021703543.1 | 9 - 10 | 2676 | 95 | ||
XM_021847852.1 | XP_021703544.1 | 9 - 10 | 2676 | 95 |
