Assay Details
Gene Symbol: LOC5567991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847846.1 | XP_021703538.1 | 8 - 9 | 1897 | 57 | |
XM_021847847.1 | XP_021703539.1 | 8 - 9 | 1891 | 57 | ||
XM_021847848.1 | XP_021703540.1 | 8 - 9 | 1634 | 57 | ||
XM_021847849.1 | XP_021703541.1 | 8 - 9 | 1523 | 57 | ||
XM_021847850.1 | XP_021703542.1 | 12 - 13 | 3253 | 57 | ||
XM_021847851.1 | XP_021703543.1 | 11 - 12 | 3097 | 57 | ||
XM_021847853.1 | XP_021703545.1 | 2 - 3 | 1340 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021847853.1 transcripts.
Set Membership: |
![]() ![]() |
