Assay Details
Gene Symbol: LOC5579975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841705.1 | XP_021697397.1 | 4 - 5 | 1133 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5579975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841705.1 | XP_021697397.1 | 4 - 5 | 1133 | 91 |
Set Membership: |
![]() ![]() ![]() |