Assay Details
Gene Symbol: LOC5575240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655595.2 | XP_001655645.1 | 2 - 3 | 355 | 68 | |
XM_021841844.1 | XP_021697536.1 | 1 - 2 | 183 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
