Assay Details
Gene Symbol: LOC5565620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844846.1 | XP_021700538.1 | 1 - 2 | 601 | 82 | |
XM_021844847.1 | XP_021700539.1 | 1 - 2 | 601 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021844846.1, XM_021844847.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
