Assay Details
Gene Symbol: LOC5565620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844846.1 | XP_021700538.1 | 15 - 16 | 8604 | 57 | |
XM_021844847.1 | XP_021700539.1 | 13 - 14 | 7155 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
