Assay Details
Gene Symbol: LOC5573302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842953.1 | XP_021698645.1 | 3 - 4 | 1405 | 85 | |
XM_021842954.1 | XP_021698646.1 | 3 - 4 | 1280 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
