Assay Details
Gene Symbol: LOC5570561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853210.1 | XP_021708902.1 | 3 - 4 | 1883 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021853210.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
