Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 12 - 13 | 2966 | 67 | |
XM_021843936.1 | XP_021699628.1 | 12 - 13 | 2961 | 67 | ||
XM_021843937.1 | XP_021699629.1 | 12 - 13 | 2948 | 67 | ||
XM_021843938.1 | XP_021699630.1 | 12 - 13 | 2943 | 67 | ||
XM_021843939.1 | XP_021699631.1 | 11 - 12 | 2429 | 67 | ||
XM_021843940.1 | XP_021699632.1 | 11 - 12 | 2459 | 67 | ||
XM_021843941.1 | XP_021699633.1 | 9 - 10 | 1566 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
