Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 16 - 17 | 4104 | 118 | |
XM_021843936.1 | XP_021699628.1 | 16 - 17 | 4099 | 118 | ||
XM_021843937.1 | XP_021699629.1 | 16 - 17 | 4086 | 118 | ||
XM_021843938.1 | XP_021699630.1 | 16 - 17 | 4081 | 118 | ||
XM_021843939.1 | XP_021699631.1 | 15 - 16 | 3567 | 118 | ||
XM_021843940.1 | XP_021699632.1 | 15 - 16 | 3597 | 118 | ||
XM_021843941.1 | XP_021699633.1 | 13 - 14 | 2704 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
