Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 3 - 4 | 1113 | 61 | |
XM_021843936.1 | XP_021699628.1 | 3 - 4 | 1108 | 61 | ||
XM_021843937.1 | XP_021699629.1 | 3 - 4 | 1095 | 61 | ||
XM_021843938.1 | XP_021699630.1 | 3 - 4 | 1090 | 61 | ||
XM_021843939.1 | XP_021699631.1 | 2 - 3 | 576 | 61 | ||
XM_021843940.1 | XP_021699632.1 | 2 - 3 | 606 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
