Assay Details
Gene Symbol: LOC5574477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843935.1 | XP_021699627.1 | 8 - 9 | 2255 | 92 | |
XM_021843936.1 | XP_021699628.1 | 8 - 9 | 2250 | 92 | ||
XM_021843937.1 | XP_021699629.1 | 8 - 9 | 2237 | 92 | ||
XM_021843938.1 | XP_021699630.1 | 8 - 9 | 2232 | 92 | ||
XM_021843939.1 | XP_021699631.1 | 7 - 8 | 1718 | 92 | ||
XM_021843940.1 | XP_021699632.1 | 7 - 8 | 1748 | 92 | ||
XM_021843941.1 | XP_021699633.1 | 5 - 6 | 855 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
