Assay Details
Gene Symbol: LOC5570780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659321.2 | XP_001659371.1 | 4 - 5 | 828 | 75 | |
XM_021853156.1 | XP_021708848.1 | 4 - 5 | 788 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
