Assay Details
Gene Symbol: LOC5565457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649716.2 | XP_001649766.2 | 1 - 2 | 450 | 75 | |
XM_021854872.1 | XP_021710564.1 | 2 - 3 | 508 | 75 | ||
XM_021854873.1 | XP_021710565.1 | 2 - 3 | 447 | 75 | ||
XM_021854874.1 | XP_021710566.1 | 2 - 3 | 518 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
