Assay Details
Gene Symbol: LOC5571092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850999.1 | XP_021706691.1 | 2 - 3 | 1356 | 101 | |
XM_021851000.1 | XP_021706692.1 | 2 - 3 | 1278 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
