Assay Details
Gene Symbol: LOC5578123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656770.2 | XP_001656820.1 | 3 | 632 | 104 | |
GenBank mRNA | AY432101.1 | - | 1 | 222 | 106 | |
AY552053.1 | - | 3 | 533 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
