Assay Details
Gene Symbol: LOC5564541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648840.2 | XP_001648890.1 | 10 - 11 | 2106 | 74 | |
XM_021846957.1 | XP_021702649.1 | 9 - 10 | 1890 | 74 | ||
GenBank mRNA | AY432103.1 | - | 1 - 2 | 96 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
