Assay Details
Gene Symbol: LOC5580263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854751.1 | XP_021710443.1 | 3 - 4 | 1711 | 68 | |
XM_021854752.1 | XP_021710444.1 | 3 - 4 | 1692 | 68 | ||
XM_021854753.1 | XP_021710445.1 | 3 - 4 | 1598 | 68 | ||
XM_021854754.1 | XP_021710446.1 | 3 - 4 | 1668 | 68 | ||
XM_021854755.1 | XP_021710447.1 | 3 - 4 | 1529 | 68 | ||
XM_021854756.1 | XP_021710448.1 | 3 - 4 | 1502 | 68 | ||
XM_021854757.1 | XP_021710449.1 | 3 - 4 | 1424 | 68 | ||
XM_021854759.1 | XP_021710451.1 | 3 - 4 | 1342 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
