Assay Details
Gene Symbol: LOC5563881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855227.1 | XP_021710919.1 | 7 - 8 | 2142 | 77 | |
XM_021855228.1 | XP_021710920.1 | 7 - 8 | 2142 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
