Assay Details
Gene Symbol: LOC5570796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653235.2 | XP_001653285.1 | 4 - 5 | 1020 | 67 | |
XM_021842898.1 | XP_021698590.1 | 4 - 5 | 984 | 67 | ||
XM_021842899.1 | XP_021698591.1 | 4 - 5 | 792 | 67 |

More Information
Set Membership: |
![]() ![]() |
