Assay Details
Gene Symbol: LOC5570796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653235.2 | XP_001653285.1 | 7 - 8 | 1746 | 65 | |
XM_021842898.1 | XP_021698590.1 | 7 - 8 | 1710 | 65 | ||
XM_021842899.1 | XP_021698591.1 | 7 - 8 | 1518 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
