Assay Details
Gene Symbol: LOC5569016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652624.2 | XP_001652674.1 | 2 - 3 | 396 | 85 | |
GenBank mRNA | AY432125.1 | - | 2 - 3 | 304 | 85 | |
DQ440189.1 | - | 2 - 3 | 214 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
