Assay Details
Gene Symbol: LOC5569016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652624.2 | XP_001652674.1 | 4 - 5 | 734 | 97 | |
GenBank mRNA | AY432125.1 | - | 4 - 5 | 642 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
